To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG1, lambda Isotype Ctrl
- Clonality:Monoclonal
- Clone:G0114F7
- Host:Rat
- Isotype:IgG1 lambda
- Reactivity:Rat
- Antigen symbol:IgG1 lambda
- Size:500 µg
- Form:0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C. This Ultra-LEAF™ solution contains no preservative; handle under aseptic conditions.
- Concentration:1.00 mg/mL
- Shipping temperature:Ambient RT
- Purification:The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Cat. no.:CA10761-250
- Supplier No.:401916
Specifications
About this item
Ultra-LEAF™ Purified Rat IgG1, λ Isotype Ctrl [G0114F7]; Isotype: Rat IgG1, λ; Reactivity: Rat; Apps: FC; Size: 500 μg
Each lot of this rat IgG1, λ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG1 lambda Isotype Ctrl
Clonality: Monoclonal
Clone: G0114F7
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1 lambda
Reactivity: Rat