About this item
Purified Rat IgG1, λ Isotype Ctrl [G0114F7]; Isotype: Rat IgG1, λ; Reactivity: Rat; Apps: FC; Size: 500 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG1 lambda Isotype Ctrl
Clonality: Monoclonal
Clone: G0114F7
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1 lambda
Reactivity: Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10761-228
- CA10761-230
- Antigen name:
- IgG1, lambda Isotype Ctrl
- IgG1, lambda Isotype Ctrl
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- G0114F7
- G0114F7
- Reactivity:
- Rat
- Rat
- Host:
- Rat
- Rat
- Isotype:
- IgG1 lambda
- IgG1 lambda