About this item
Alexa Fluor® 700 Mouse IgG1, κ Isotype Ctrl (ICFC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 tests
This reagent is developed for intracellular immunofluorescent staining for flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. It is highly recommended that the reagent be titrated for optimal performance for each application.
Type: Control
Antigen: IgG1, k isotype ctrl (ICFC)
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10761-094
- CA10761-096
- Antigen name:
- IgG1, k isotype ctrl (ICFC)
- IgG1, k isotype ctrl (ICFC)
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MOPC-21
- MOPC-21
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa