To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GM-CSF
- Clonality:Monoclonal
- Clone:BVD2-21C11
- Gel shift:Yes
- Gene ID:1437
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Antigen symbol:GM-CSF
- ELISA:Yes
- Size:100 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and EDTA.
- Antigen synonyms:Eo-CSF|CSF-α|BPA|CSF2|Granulocyte/macrophage colony-stimulating factor|Pluripoietin-α
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:1.00 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:<I>E. coli</I>-expressed, recombinant human GM-CSF.
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10761-616
- Supplier No.:502315
Specifications
About this item
Purified anti-human GM-CSF (MaxPar® Ready) [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus, Cynomolgus; Apps: ELISA Capture, CyTOF®; Size: 100 μg
Each lot of this antibody is quality control tested by ELISA assay. This product is suitable for use with the MaxPar® Metal Labeling Kits. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the MaxPar® antibody labeling protocol). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol. For ELISA application, a concentration range of 1-4 μg/ml (BVD2-21C11) is recommended. The purified BVD2-21C11 antibody is useful as the capture antibody in a sandwich ELISA assay, when used in conjunction with the biotinylated BVD2-23B6 antibody as the detecting antibody.
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: BVD2-21C11
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Cross-Reactivity: Rhesus;Cynomolgus