Orthophosphoric acid ≥85-88% (by acidimetry) NF, ACS, J.T.Baker®
Catalog #: CAJT0240-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7664-38-2
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE drum, black
- Size:318 kg (700 lbs.)
- Cat. no.:CAJT0240-7
Specifications
Specification Test Results
- Meets N.F. Requirements:
- GMP Manufactured Product:
- Meets ACS Reagent Chemical Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (H₃PO₄):85.0 - 88.0 %
- NF - Identification:Passes Test
- NF - Nitrate (NO₃):Passes Test
- NF - Phosphorous or Hypophosphorous Acid:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Alkali Phosphates:Passes Test
- Assay (H₃PO₄) (by acidimetry):85.0 - 87.0 %
- Calcium (Ca):≤0.002 %
- Chloride (Cl):≤3 ppm
- Color (APHA):≤10
- Heavy Metals (as Pb):≤8 ppm
- Insoluble Matter:≤0.001 %
- ACS - Magnesium (Mg):≤0.002 %
- Nitrate (NO₃):≤5 ppm
- Reducing Substances:Passes Test
- Sulfate (SO₄):≤12 ppm
- Trace Impurities - Antimony (Sb):≤20.000 ppm
- Trace Impurities - Arsenic (As):≤0.500 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
- Trace Impurities - Manganese (Mn):≤0.500 ppm
- Trace Impurities - Potassium (K):≤40.000 ppm
- Trace Impurities - Sodium (Na):≤200.000 ppm
- Volatile Acids (as CH₃COOH):≤0.001 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232) - Information on elemental impurities for:
- this product is available on the associated Product Regulatory Data:
- Sheet and elemental impurity profile report.:
- J.T.Baker®, Orthophosphoric acid ≥85-88% (by acidimetry) NF, ACS:
Specification Test Results
About this item
Synonyms: Phosphoric acid
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85-88
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: ~ 21 °C
:Flash Pt:
:Flash Pt:
:M.W. 98 g/mol
:21
:158
UN:1805
:8
:III
:GHS05
:GHS07
MW:98 g/mol
Storage Temperature:Ambient
:M.O.C. Category R
:12,07500
:14,07344
Formula:H₃PO₄
MDL Number:MFCD00011340
CAS Number:7664-38-2
Customers Who Bought This Also Bought