DuPont™ Proshield® 10 Lab Coats
Supplier: DuPont
Certificates
About this item
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
- Laydown collar
- Open wrists
- Length extends below hips
- Front snap closure
- Two pockets
- Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Lab coat offers front snap closures for easy donning and doffing. Additional features include laydown collar, open wrists, and two front pockets for holding writing instruments, small tools, or parts.
Certifications: Thickness meets ASTM D1117.
Specifications
- Catalog No:
- 10016-288
- 10016-774
- 10016-276
- 10016-278
- 10016-280
- 10016-776
- 10016-778
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory