About this item
Analyze oxygen, nitrogen, methane, carbon monoxide, and carbon dioxide with one ShinCarbon ST column at room temperature.
- Separate permanent gases, including carbon monoxide and carbon dioxide, without cryogenic cooling
- Rapid separations of permanent gas/light hydrocarbon mixtures
- Excellent compatibility with most GC detectors—minimal bleed, minimal baseline rise.
- Preconditioned, less than 30 minutes to stabilize
- Maximum temperature of 280/300 °C
- SilcoSmooth® stainless steel
The rapid, above-ambient analyses these columns provide is a great convenience.
ShinCarbon ST is a highly stable material. Its 300 °C upper programmed temperature limit minimizes bleed and baseline rise during temperature programming, making the material compatible with most detection systems used for gas analysis, including TCD or HID. All ShinCarbon ST columns are fully conditioned in an oxygen/moisture-free environment for your convenience. This minimizes stabilization time (less than 30 minutes) when installing a new column which, in turn, reduces downtime.
Excellent thermal stability of the high surface area carbon, combined with careful conditioning during column manufacturing, ensures low-bleed operation and rapid stabilization when installing a new column.
Specifications
- Catalog No:
- 10847-846
- 10848-628
- 10847-844
- 10847-842
- 10848-620
- 10847-810
- 10847-826
- 10803-502
- 10848-634
- Description:
- ShinCarbon ST Micropacked Column, SilcoSmooth Tubing
- ShinCarbon ST Packed Column, SilcoSmooth Tubing, Varian Configuration
- ShinCarbon ST Micropacked Column, SilcoSmooth Tubing
- ShinCarbon ST Micropacked Column, SilcoSmooth Tubing
- ShinCarbon ST Packed Column, SilcoSmooth Tubing, General Configuration
- ShinCarbon ST Micropacked Column, SilcoSmooth Tubing
- ShinCarbon ST Micropacked Column, SilcoSmooth Tubing
- ShinCarbon ST Packed Column, SilcoSmooth Tubing, Agilent Configuration
- ShinCarbon ST Packed Column, SilcoSmooth Tubing, PE Auto Sys Configuration
- Temperature limit:
- up to 300 °C
- up to 300 °C
- up to 300 °C
- up to 300 °C
- up to 300 °C