Rtx®-65TG Columns (Fused Silica), Restek
Supplier: Restek
Certificates
About this item
The Rtx®-65TG phase resolves triglycerides by degree of unsaturation as well as by carbon number.
- Application-specific columns, specially tested for triglycerides
- Stable to 370 °C
- High-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
Because of the chemistry required to achieve 370 °C thermal stability, an Rtx®-65TG column should not be used for the analyses of polar compounds.
Specifications
- Catalog No:
- RK17005
- RK17009
- RK17010
- RK17006
- RK17008
- Description:
- Rtx-65TG
- Rtx-65TG
- Rtx-65TG
- Rtx-65TG
- Rtx-65TG
- Temperature limit:
- 40 to 370 °C
- 40 to 370 °C
- 40 to 370 °C
- 40 to 370 °C
- 40 to 370 °C