MXT®-50 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
Certificates
About this item
MXT®-50 columns are good for general-purpose applications involving pesticides, herbicides, pharmaceuticals, rosin acids, phthalate esters, and sterols.
- General-purpose columns for pesticides, herbicides, rosin acids, phthalate esters, and sterols
- Temperature range: 0 to 300 °C
- Equivalent to USP G3 phase
- 4.5" standard coil diameter
- Mid-polarity phase; Crossbond® phenyl methyl polysiloxane
These columns are constructed of Siltek®-treated stainless steel.
Specifications
- Catalog No:
- RK70569
- 10851-686
- 10851-688
- Description:
- MXT-50
- MXT-50
- MXT-50
- Temperature limit:
- 0 to 280/300 °C
- 0 to 260/280 °C
- 0 to 260/280 °C