MXT®-20 Columns (Siltek®-treated stainless steel), Restek
Catalog #: 10851-668
Supplier: Restek
Specifications
- Column Format:Capillary
- Film thickness:0.25 µm
- I.D.:0.25 mm
- Length:15 m
- Description:MXT-20
- Temperature limit:-20 to 300/340 °C
- Cat. no.:10851-668
- Packing Material:Siltek®-treated stainless steel
- Supplier No.:70320
Specifications
About this item
MXT®-20 polymer is synthesized to exacting standards so that all residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight mono-modal distribution and extremely low bleed.
- General-purpose columns for volatile compounds, flavor compounds, and alcoholic beverages
- Temperature range: −20 to 340 °C
- Equivalent to USP G28, G32 phases
- Low to mid-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
MXT®-20 columns are made of Siltek®-treated stainless steel and feature extremely low bleed levels.
Customers Who Bought This Also Bought