Rt®-γDEXsa Columns (Fused Silica), Restek
Supplier: Restek
Certificates
About this item
By adding β or γ cyclodextrin to Restek's bonded Rtx®-1701 stationary phase, we greatly enhance overall utility and column lifetime for Restek's chiral columns, compared to columns that have pure cyclodextrin stationary phases.
- 2,3-di-acetoxy-6-O-tert-butyl dimethylsilyl gamma cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane
- Improved column lifetime
- Enhanced column utility
Separations of more than one hundred chiral compounds have been achieved using Restek's unique DEX columns, and Restek's columns continue to demonstrate stability after hundreds of temperature program cycles.
Uses: Larger organic molecules. Also useful for flavor compounds in fruit juices.
Specifications
- Catalog No:
- 10849-356
- 10849-354
- Description:
- Rt-γDEXsa
- Rt-γDEXsa
- Temperature limit:
- 40 to 230 °C
- 40 to 230 °C