MXT®-5 Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
Certificates
About this item
All residual catalysts and low molecular weight fragments are removed from the MXT®-5 polymer, providing a tight mono-modal distribution and extremely low bleed.
- General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, and semi-volatiles
- Equivalent to USP G27, G36 phases
- 4.5" standard coil diameter
- Low-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
The diphenyl dimethyl polysiloxane stationary phase is the most popular GC stationary phase and is used in a wide variety of applications.
Specifications
- Catalog No:
- RK70239
- RK70224
- 10851-838
- 10851-794
- 10851-796
- 10851-798
- 10851-864
- 10851-866
- 10851-868
- 10851-870
- 10851-872
- RK70240
- 10851-800
- 10851-802
- 10851-804
- 10851-806
- 10851-808
- 10851-810
- 10851-812
- 10851-088
- RK70236
- 10851-840
- 10851-818
- 10851-844
- 10851-846
- 10059-582
- 10851-848
- 10851-850
- 10851-852
- 10059-580
- 10851-822
- 10851-824
- 10851-826
- 10851-828
- 10851-830
- 10851-832
- 10851-834
- 10851-836
- 10851-820
- Description:
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- MXT-5
- Temperature limit:
- -60 to 330/400 °C
- -60 to 360/430 °C
- -60 to 270/360 °C
- -60 to 360/430 °C
- -60 to 340/430 °C
- -60 to 340/430 °C
- -60 to 325/430 °C
- -60 to 325/400 °C
- -60 to 325/430 °C
- -60 to 315/400 °C
- -60 to 315/400 °C
- -60 to 330/400 °C
- -60 to 340/430 °C
- -60 to 330/430 °C
- -60 to 340/430 °C
- -60 to 340/430 °C
- -60 to 330/360 °C
- -60 to 330/400 °C
- -60 to 315/400 °C
- -60 to 360/430 °C
- -60 to 315/400 °C
- -60 to 270/360 °C
- -60 to 310/340 °C
- -60 to 290/360 °C
- -60 to 270/360 °C
- -60 to 330/430 °C
- -60 to 290/360 °C
- -60 to 290/360 °C
- -60 to 290/360 °C
- -60 to 330/430 °C
- -60 to 310/360 °C
- -60 to 310/340 °C
- -60 to 325/360 °C
- -60 to 325/360 °C
- -60 to 325/360 °C
- -60 to 310/360 °C
- -60 to 300/360 °C
- -60 to 300/360 °C
- -60 to 310/360 °C