Rtx®-200MS Low-Bleed GC-MS Columns (fused silica), Restek
Supplier: Restek
Certificates
About this item
Rtx®-200 columns have accomplished many difficult separations not possible on any other bonded stationary phase.
- General-purpose columns
- Equivalent to USP G6 phase
- Inert properties
- Mid-polarity columns
- Low bleed
- Exceptional thermal stability
The trifluoropropylmethyl polysiloxane stationary phase has a unique selectivity that changes elution orders and resolves compounds that phenyl, cyano, or Carbowax® phases cannot. The Rtx®-200 column offers exceptional thermal stability, low bleed, and superior inertness—even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs. Column specifically tested for low-bleed performance.
Specifications
- Catalog No:
- 10058-498
- 10850-046
- 10058-500
- 10058-494
- 10058-496
- 10850-060
- 10851-992
- 10851-976
- 10850-054
- Description:
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Rtx-200MS
- Temperature limit:
- -20 to 290/310 °C
- -20 to 320/340 °C
- -20 to 290/310 °C
- -20 to 320/340 °C
- -20 to 310/330 °C
- -20 to 310/330 °C
- -20 to 320/340 °C