About this item
These guard columns and transfer lines are designed to prevent water vapor from reaching the surface, preventing aggressive hydrolysis.
- Extend analytical column lifetime by preventing degradation from harsh 'steam-cleaning' water injections
- Maximum temperature: 325 °C
- 30-meter lengths are banded in 5-meter sections
- Diameters greater than 0.10 mm are tested with the Grob test mix to ensure high inertness
When transfer lines from purge-and-trap systems, air monitoring equipment, or other instruments carry condensed water vapor, deactivated column tubing quickly becomes active because of the creation of free silanol groups.
With Hydroguard® deactivated tubing, a unique deactivation chemistry creates a high-density surface that is not readily attacked by aggressive hydrolysis. The high-density surface coverage of the Hydroguard® deactivation layer effectively prevents water vapor from reaching the surface beneath. Use Hydroguard® tubing for connecting GCs to these devices:
Headspace analyzers
Air analysis equipment and concentrator units
Purge-and-trap systems
These silanol groups adsorb active oxygenated compounds, such as alcohols and diols.