Rt®-βDEXm Columns (fused silica), Restek
Supplier: Restek
Certificates
About this item
By adding β or γ cyclodextrin to Restek's bonded Rtx®-1701 stationary phase, we greatly enhance overall utility and column lifetime for Restek's chiral columns, compared to columns that have pure cyclodextrin stationary phases.
- Improve overall utility
- Longer column lifetime
- Ensured stability
Separations of more than one hundred chiral compounds have been achieved using Restek's unique DEX columns, and Restek's columns continue to demonstrate stability after hundreds of temperature program cycles.
Uses: General-purpose chiral phase with many published applications.
Permethylated beta cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane.
Specifications
- Catalog No:
- RK13101
- RK13100
- Description:
- Rt-βDEXm
- Rt-βDEXm
- Temperature limit:
- 40 to 230 °C
- 40 to 230 °C