Specifications
- Capacity:59 ml (2 oz.)
- Description:Medicine Cups
- Supplier No.:96-7027
Specifications
About this item
Sterile, disposable transparent plastic sample cups for medicinal purposes.
- Latex free
- Graduated
- Premium OR-Grade
Sklar's® Medicine Cup is used for dispensing both liquid and dry medications. The cups are made of polypropylene and are latex-free. They have easy to read markings on the interior surface