About this item
Sklar's Heaney Uterine Curette is used for tissue removal from the uterus.
It has a straight, malleable shaft that can be manipulated to obtain an adequate tissue sample or to remove all tissue as necessary. The tip is a serrated loop. The handle is flat and gradually tapers to become very narrow until it reaches the loop tip.