Q-sep® Bottle Top Solvent Dispenser, Restek
Catalog #: 10015-344
Supplier: Restek
Specifications
- Description:Q-sep bottle top Solvent dispenser, 2.5 ml–30 ml
- Cat. no.:10015-344
- Supplier No.:23990
Specifications
About this item
Accurately and precisely dispense liquids for QuEChERS extractions with this versatile pump.
- Base features 30 mm threads and includes four adapters (25 mm, 28 mm, 38 mm, and 45 mm)
- Individually calibrated in accordance with ISO 8655 standards (certificate included) and can also be recalibrated by the user
- PTFE, glass, and polypropylene construction for excellent chemical compatibility and 100% autoclavability
- Integral safety discharge reduces risk of accidental dispensing, and nozzle cap prevents dripping
- Easy to disassemble for cleaning and servicing
- SPE accessory
A quick, simple adjustment lets you set the output volume anywhere from 2.5 mL to 30 mL per stroke, and the included adaptors will accommodate most reagent bottles. Adjustment knob offers 56 output volume settings from 2.5 mL to 30 mL per stroke (0.5 mL increments)—ideal for QuEChERS methods.
Customers Who Bought This Also Bought