About this item
Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine high affinity for biotinylated target molecules with very low non specific binding. Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine fast reaction kinetics with very low non specific binding for increased throughput and precision. Applications include automated immunoassays, immunoprecipitation, and protein purification. Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation. Sera-Mag SpeedBeads and Sera-Mag Streptavidin-Coated Magnetic Particles provide a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules.
- Non-surfactant, non-protein blocking reagent provides very low non-specific binding and high binding capacity for biotinylated target molecules
- Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
- Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
- Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10146-022
- CA10146-018
- CA10146-020
- Binding capacity:
- 3000 - 4000 pmole/mg beads
- 3000 - 4000 pmole/mg beads
- 3000 - 4000 pmole/mg beads
- Particle size:
- nominal 1 µm Ø
- nominal 1 µm Ø
- nominal 1 µm Ø
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Next-Generation Sequencing (NGS) Solutions