Specifications
- Length:31.7 cm (121/2")
- Description:Merit™ Sims Uterine Sound, Plain
- Material:Silver-Plated Stainless Steel
- Cat. no.:10185-324
- Supplier No.:98-384
Specifications
About this item
The Merit™ Sims Uterine Sound is a malleable, silver-plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal.
It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features three vertical grooves for improved grip.