MicroSeal SuperSorb® Lite Cleanroom Laundered Knit Wiper, Berkshire
Supplier: BERKSHIRE CORPORATION MX
Certificates
About this item
MicroSeal SuperSorb® Lite are high absorbency 100% continuous filament polyester knit wiper.
Its design provides texture and extra durability, when cleaning stubborn residues. It is an excellent wiper for applications where high concentrations of liquid are in use, as well as cleaning and wiping rough surfaces.
Ultrasonically sealed edges for spill control in processing areas, where absorbency is important.
Ordering information: Quality control and lot traceability identification on each case.
Customers Who Bought This Also Bought