KoolTemp® GTS-218 Pre-Qualified Shippers, Cold Chain Technologies
Supplier: Cold Chain Technologies
Certificates
About this item
Simple universal pack-out for year-round performance in extreme temperatures.
- Pre-qualified for Multiple Temperature Ranges
- Refrigerated (2-8°C) for 72 hours
- Room Temperature (15–25°C or 10–30°C) for 96 to 120 hours
- Frozen (below -60°C, -40°C, and -20°C) for up to 120+ hours
Cost effective for single use shipments but also robust enough for return/re-use. Made from environmentally safe and reusable materials.
Refrigerant: Refrigerated and Room Temp: (12) Koolit 338F Bricks (8) Koolit 339F Bricks; Frozen: 53 lbs. of Pelletized Dry Ice [Refrigerant Used in Solution]
Ordering information: Model GTS-218-KD is shipped to the customer with the refrigerants in a separate box from the shipper.
Customers Who Bought This Also Bought