ESD Modular Trays, Quantum Storage Systems
Supplier: QUANTUM STORAGE SYSTEMS
Certificates
About this item
These trays are ESD and comply with ANSI/ESD standards for the use in PC board assembly and handling.
- Withstand temperatures of −40 °F to +350 °F
- Trays are resistant to abrasion and chemicals
- Trays are permanently conductive, properties are unaffected by washing in steam or water
All trays are manufactured of fiberglass reinforced thermoset material, allowing up to 200 lb. weight capacity when evenly distributed.
Customers Who Bought This Also Bought