Polyurethane Foam Sock Swabs, Berkshire
Supplier: Berkshire
Certificates
About this item
These general purpose swabs are designed for cleaning small and confined areas.
- Low particles, fibers, ions, and extractables
- Good chemical resistance
- Soft and non-abrasive with no contaminating adhesives
- Good sorbency
- Clean room class 100 to 100000
Its laundered open cell foam head combines superior cleanliness with particle entrapment capability.
Ordering information: 100 swabs per pack, 10 packs per case (double packed).
Customers Who Bought This Also Bought