About this item
Future-proof any laboratory with the SpectraMax® Paradigm® Multi-Mode Microplate Reader.
- High-speed detection
- Excellent sensitivity and performance
- Applications at user fingertips
- Array of detection cartridge capabilities
- Automatic Z-height optimization
- Patented LED intensity adjustment
- Up to 1536-well format
- Temperature control
Get the system that delivers the data needed today, and when additional requirements arise, transform the reader with a detection-mode addition upgrade that takes less than two minutes
Flexible design allows for single or dual excitation and emission, to address existing needs and future expansion. User upgradable microplate reader in less than 2 minutes. Features automatic Z-height optimization (both top- and bottom-read configurations). Fluorescence measurements no longer require users to adjust the gain. TUNE Cartridge available for unmatched sensitivity in wavelength tunable detection. Custom Cartridge development available to meet specific research needs. SpectraDrop™ Plate protocols enabling micro volume applications measurement are included.
The SpectraMax® Paradigm® reader can perform absorbance, fluorescence, time-resolved fluorescence (including HTRF), fluorescence polarization, AlphaScreen®, AlphaLISA®, and luminescence measurements. Detection cartridge modularity allows users to configure the system to meet current needs, while providing the flexibility to address future applications.
Specifications
- Catalog No:
- 10822-512
- CA10822-512
- Light source:
- High powered LED/flash lamp
- High powered LED/flash lamp
- Wavelength:
- Photometry
- Photometry
- Plate format:
- 6- to 1536-well
- 6- to 1536-well
- Shaking:
- Linear, orbital
- Linear, orbital
- Temperature uniformity:
- ±0.75 °C
- ±0.75 °C