Equivalence C18, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
- Excellent efficiency and reproducibility
- Available in 3 or 5 µm particle sizes
Maximum recommended pH: 2 to 8.