ContactUsLinkComponent
BD Vacutainer® Critical Care Syringes, BD Medical
BD Vacutainer® Critical Care Syringes, BD Medical
Catalog #: CABD364356
Supplier:  BD
SYR ABG ALINE 1(.6) S/T CE
BD Vacutainer® Critical Care Syringes, BD Medical
Catalog #: CABD364356
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    1 ml
  • Description:
    BD A-Line™ critical care syringes, without needle, slip tip, with 30 units of spray dried heparin
  • Supplier No.:
    364356
Specifications
About this item
BD Vacutainer® critical care syringes can be used to measure arterial blood gas, ionized calcium, ionized magnesium, as well a perform critical care tests
BD A-Line™ syringes contain calcium balanced lithium heparin spray dried on the sides of the syringes.
Customers Who Bought This Also Bought