Flame Resistant Work Shirt with Max Comfort™ Material, Gray, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
- Comfortable 88% cotton/12% nylon blend
- Two chest pockets
- NFPA 2112, NFPA 70E, UL Certified
The S1 Work Shirt features a comfortable 88% cotton/12% nylon blend that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.
Specifications
- Catalog No:
- 10813-400
- 10813-398
- 10813-572
- 10813-396
- 10813-404
- 10813-566
- 10813-394
- 10813-570
- 10813-392
- 10813-568
- 10813-390
- Model:
- S1
- S1
- S1
- S1
- S1
- S1
- S1
- S1
- S1
- S1
- S1
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- 88% cotton/12% nylon
- Thickness:
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
- 6.5 oz.
Specifications
Related Information
Customers Who Bought This Also Bought