DuPont™ ProShield® 70 Boot Covers with Skid Resistant Sole
Catalog #: 10016-146
Supplier: DuPont
Specifications
- Garment application:Laboratory
- Length:171/2"
- Shoe size:S
- Color:Gray
- Height:10"
- Cat. no.:10016-146
- Supplier No.:P3452SGY0000
Specifications
About this item
ProShield® 70 fabric, used exclusively for shoe and boot cover products, is a spunbonded polypropylene substrate that is coated with a low-density polyethylene film for enhanced skid resistance and barrier performance.
- Skid-resistant sole
- Coverage up to calf
- Serged seams
- One size fits most
These boot covers feature a ProShield® 70 skid-resistant sole for enhanced skid resistance and coverage up to the calf.
Customers Who Bought This Also Bought