Pivoting Blade Impeller, 2.4", Caframo
Supplier: Caframo
Certificates
About this item
The Caframo Pivoting Blade Impeller features a 2.4" diameter, and is pinned to a 5/16" by 16" shaft
The impeller will vortex 5 gallons of water at 800rpm. Designed for use with narrow neck vessels.
Composed of 316L electropolished stainless steel, it offers moderate shear with moderate vortexing characteristics.
Customers Who Bought This Also Bought