Crossed Blade Impeller, 2", Caframo
Supplier: Caframo
Certificates
About this item
The Caframo Crossed Blade Impeller features a 2" diameter, and is welded to a 5/16" diameter by 16" shaft
The impeller will vortex 5 gallons of water at 900rpm. Radial flow.
Composed of 316L electropolished stainless steel, it offers very high shear with high vortexing characteristics.
Customers Who Bought This Also Bought