Silica Gel GHLF UNIPLATES™ TLC Plates, Analtech, Miles Scientific
Supplier: Analtech
Certificates
About this item
These plates are 100% water resistant and are compatible with aqueous developing solvents for efficient separations.
- Contains no organic materials
- Suitable for aggressive visualization procedures
- Tough writable surface
They are less rugged than the plates with organic binder but still provide improved handling and sample application.
Specifications
- Catalog No:
- 97054-124
- 57457-162
- Average particle size:
- 15 µm
- 15 µm