Order Entry
ContactUsLinkComponent
Mueller Hinton Agar, Hardy Diagnostics
Mueller Hinton Agar, Hardy Diagnostics
Catalog #: 10054-352
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Mueller Hinton Agar
Mueller Hinton Agar, Hardy Diagnostics
Catalog #: 10054-352
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. no.:
    10054-352
  • Dimensions:
    15×100 mm
  • Volume:
    28 ml
  • Supplier No.:
    G45

Specifications

About this item

Mueller Hinton Agar is ideal for antimicrobial susceptibility testing of non-fastidious bacteria.

  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Packaged in sleeves of 10 plates in a 'breathable' celluose bag that prevents build-up of condensation and excess moisture
  • Holds up to five animicrobial discs

Ideal for antimicrobial susceptibility testing of non-fastidious bacteria by utilizing disk diffusion sensitivity method (Kirby-Bauer). It is a smaller plate and it will hold upto five antimicrobial discs.

Customers Who Bought This Also Bought