Automatic Potentiometric Titrators, Koehler Instrument
: Avantor
The Automatic Titrator can accept a variety of electrodes to cater to various applications in different fields.
- Provided with two-point auto calibration and standardization (zero offset)
- Capable of displaying pH and mV of the sample, with temperature compensation
- Supplied with high speed vortex stirrer with digital speed indication
- Specially designed stirrer provides excellent homogenous mixing of samples
- Microcontroller based stirrer system
The Automatic Titrator is capable of performing a wide range of titrations like acid-base or aqueous, redox, complexometric titration or EDTA, blank, silver assay, non-aqueous, argentometric or precipitation, voltametric / KF and back titations.
Frequently Bought Together