Sample Thieves, All Levels, Koehler
Catalog #: 10118-912
Supplier: Koehler
Specifications
- Description:All-Levels Sample Thief
- Cat. no.:10118-912
- Supplier No.:K27800
Specifications
About this item
Equipped with an adjustable needle valve opening instead of a plunger to control the rate of flow during "all-levels" and "running" sampling from storage tanks.
Similar to the standard (473mL) Sample Thief (Bacon Bomb), plated brass construction.
Customers Who Bought This Also Bought