Specifications
- Length:140 mm (51/2")
- Material:Stainless Steel
- Cat. no.:10193-334
- Jaw Type:Serrated with groove
- Tip Style:Straight
- Supplier No.:40-1775
Specifications
About this item
Needle Nose Pliers are a non-ratcheted, plier handle instrument used for grasping and manipulating k-wire.
- Tips are tapered and squared off
The working surface of this instrument is serrated and features a central groove that grips the wire more securely than a smooth surface.
They are used to guide and twist the k-wire as it is used to hold fractured bones together.