About this item
SGE jumbo syringes offer convenient use as well as versatility and reliability for the handling of large volumes of gas.
- Heavy-duty acrylic barrels
- Easy access to sample for the addition of standards or removal of subsample via secondary port
- Plunger stem is removable for ease of transportation and storage
- Accuracy and reproducibility ±2% (dispensed volume)
Compatible with a range of fittings, each syringe has a 7/16" UNEF thread and is designed to be used with Luer Lock fittings and needles.
Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Do not subject jumbo syringes to temperatures in excess of 80°C.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 60361-332
- 60361-334
- 60361-336
- Length:
- 245 mm
- 245 mm
- 245 mm
- Thread:
- 7/16" UNEF
- 7/16" UNEF
- 7/16" UNEF