To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Conjugate ELISA:HRP-Streptavidin
- Reactivity:Bovine
- Target protein:IL-6
- Size:480 assays
- Cat. no.:CAPIESS0029
- Supplier No.:ESS0029
Specifications
About this item
Assay IL-6 (bovine interleukin-6) levels in bovine cell culture supernatant samples with our Thermo Scientific Bovine IL-6 ELISA Reagent Kit.
Features of the Bovine IL-6 ELISA Reagent Kit:
Target: natural and recombinant bovine interleukin-6 (IL-6)
Format: colorimetric ELISA (TMB substrate) for 96-well microplates
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 78 to 5000 pg/mL
Sensitivity: undetermined
Sample Types: cell culture supernatant
Total Assay Time: 3.5 hours
Sample Size: 100 µL
Specificity: no significant cross-reaction with recombinant bovine IL-1β, IL-2, IL-4, IL-8, IFNγ, or TNFα.
This bovine IL-6 ELISA reagent kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. All components undergo quality control testing to minimize lot-to-lot variability.
Includes:
The Reagent Kit (previously called a "Screening Set") contains matched antibodies, standard and detection reagents but no coated plates.