Order Entry
ContactUsLinkComponent
TNF alpha ELISA Reagent Kit, Equine, Thermo Scientific
TNF alpha ELISA Reagent Kit, Equine, Thermo Scientific
Catalog #: CAPIESS0017
Supplier:  Thermo Scientific
CAS Number:  
TNF alpha ELISA Reagent Kit, Equine, Thermo Scientific
Catalog #: CAPIESS0017
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugate ELISA:
    HRP-Streptavidin
  • Target protein:
    TNF-alpha
  • Size:
    480 assays
  • Cat. no.:
    CAPIESS0017
  • Supplier No.:
    ESS0017

Specifications

About this item

Assay TNFα (equine tumor necrosis factor alpha, TNF alpha) levels in horse cell culture supernatant samples with our Thermo Scientific Equine TNF alpha ELISA Kits.

This equine TNF alpha ELISA reagent kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of horse tumor necrosis factor alpha (TNFα) in samples at 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. This economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. All components undergo quality control testing to minimize lot-to-lot variability.

Features of the Equine TNF alpha ELISA Reagent Kit:

Target: natural and recombinant horse TNFα (equine tumor necrosis factor alpha)
Format: colorimetric ELISA (TMB substrate) for 96-well microplates
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 16 to 1000 pg/mL
Sensitivity: undetermined
Sample Types: cell culture supernatant
Total Assay Time: 3 hours
Sample Size: 100 µL
Specificity: no significant cross-reaction with bovine, mouse, or rat TNFα. Partial cross-reaction occurs with human and porcine TNFα (< 10%).

Includes
The Reagent Kit (previously called a "Screening Set") contains matched antibodies, standard and detection reagents but no coated plates.

Ordering information: For Research Use Only. Not for use in diagnostic procedures. Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8˚C. See product manual for detailed contents and storage conditions for maximum stability.
Customers Who Bought This Also Bought