Order Entry
ContactUsLinkComponent
IL-6 ELISA Kit, Human, Thermo Scientific
IL-6 ELISA Kit, Human, Thermo Scientific
Catalog #: CAPIEH2IL6
Supplier:  Thermo Scientific
CAS Number:  
IL-6 ELISA Kit, Human, Thermo Scientific
Catalog #: CAPIEH2IL6
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Reactivity:
    Human
  • Target protein:
    IL-6
  • Size:
    96 assays
  • Cat. no.:
    CAPIEH2IL6
  • Supplier No.:
    EH2IL6

Specifications

About this item

The Thermo Scientific Human IL-6 ELISA Kit is used to measure IL-6 (human interleukin-6) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatants. Each kit contains one 96-well Strip Plate precoated with a human IL-6 capture antibody and sufficient detection antibody, conjugate, buffers, diluent, TMB and stop buffers, as well as lyophilized human IL-6 standard to analyze several samples.

Features of the Thermo Scientific Human IL-6 ELISA Kit: †

Target: natural and recombinant human interleukin-6 (IL-6)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 10.2 to 400 pg/mL
Sensitivity: < 1 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-10, IL-12, TNFα, IFNγ, IFNα, GM-CSF, or mouse IL-6.

Note: Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

These human IL-6 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50 to 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-6 antibody pairs and IL-6 standard only. All components undergo quality control testing to minimize lot-to-lot variability.

Ordering information: For Research Use Only. Not for use in diagnostic procedures. Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8˚C. See product manual for detailed contents and storage conditions for maximum stability.
Customers Who Bought This Also Bought