Order Entry
ContactUsLinkComponent
Pierce™ iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit, Thermo Scientific
Pierce™ iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit, Thermo Scientific
Catalog #: CAPI90103
Supplier:  Thermo Scientific
CAS Number:  
Iodotmtsixplex Isobaric Mass Tag Labeling Kit 30-reaction
Pierce™ iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit, Thermo Scientific
Catalog #: CAPI90103
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit
  • Storage conditions:
    Upon receipt store all products at -20˚C
  • Cat. no.:
    CAPI90103
  • No. of Reactions:
    30
  • Supplier No.:
    90103

Specifications

About this item

Thermo Scientific iodoTMTsixplex Isobaric Mass Tag Labeling Kit contains iodoTMT™ reagents and associated materials for the irreversible labeling of sulfhydryl groups.

  • Specifically reacts with sulfhydryl groups (reduced, unmodified cysteine residues)
  • Labeled proteins and peptides are not susceptible to reducing agents
  • Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
  • Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation)

Thermo Scientific iodoTMTsixplex Label Reagents enables enable concurrent identification and multiplexed quantitation of proteins in different samples using tandem mass spectrometry. The iodoTMT™ Reagents are sets of isobaric (mass and structure) isomers that are iodoacetyl-activated for covalent, irreversible labeling of sulfhydryl (—SH) groups. Similar to iodoacetamide, iodoTMT Reagents react specifically with reduced cysteines (Cys) in peptides and proteins. IodoTMT Reagents can be differentiated by mass spectrometry (MS), enabling quantitation of the relative abundance of cysteine modifications, such as S-nitrosylation, oxidation and disulfide bonds, in cultured cells grown or treated with different conditions.

Delivery: Kit includes 5 x 0.2 mg iodoTMT6-126 Label Reagent,, iodoTMT6-127 Label Reagent, iodoTMT6-128 Label Reagent, iodoTMT6-129 Label Reagent, iodoTMT6-130 Label Reagent, iodoTMT6-131 Label Reagent, HES Buffer (15 mL), Bond-Breaker TCEP Solution (0.5 mL), DTT No-Weigh Format (8 x 7.7 mg), Trypsin (5 x 20 µg), Trypsin Storage Solution (250 µL), Bovine Albumin (2.5 mg).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought