Order Entry
ContactUsLinkComponent
Pierce™ iodoTMTzero™ Protein Labelling Reagent, 5×0.2 mg, Thermo Scientific
Pierce™ iodoTMTzero™ Protein Labelling Reagent, 5×0.2 mg, Thermo Scientific
Catalog #: CAPI90100
Supplier:  Thermo Scientific
CAS Number:  
Iodotmtzero 5 Label Reagent 0.2Mg 5-reaction
Pierce™ iodoTMTzero™ Protein Labelling Reagent, 5×0.2 mg, Thermo Scientific
Catalog #: CAPI90100
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Description:
    iodoTMTzero™ label reagent, 5×0.2 mg
  • Size:
    5 reactions
  • Storage conditions:
    Store at -20˚C.
  • Cat. no.:
    CAPI90100
  • Supplier No.:
    90100

Specifications

About this item

The iodoTMTzero label reagent can be used as an isotopic 'light' duplex tag for MS-level protein quantitation.

  • Only reacts with sulfhydryl groups (reduced, unmodified cysteine residues), thus making it specific
  • Labeled proteins and peptides are not susceptible to reducing agents
  • Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
  • Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation) making it versatile

This tag, along with the iodoTMTsixplex™ reagents can enable the quantitation of protein expression changes in cell-based and tissue samples that may not be amenable to metabolic isotopic labeling strategies (eg. SILAC).

Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought