L-Arginine-HCl, [¹³C₆], lyophilized powder for SILAC
Catalog #: CAPI88433
Supplier: Thermo Scientific
CAS Number: 1119-34-2
Specifications
- Size:500 mg
- Cat. no.:CAPI88433
- Supplier No.:88433
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. N/A N/A
Storage Temperature:Refrigerator
CAS Number:1119-34-2
Customers Who Bought This Also Bought