L(-)-Proline, lyophilized powder for SILAC
Catalog #: CAPI88211
Supplier: Thermo Scientific
CAS Number: 147-85-3
Specifications
- Size:115 mg
- Cat. no.:CAPI88211
- Supplier No.:88211
Specifications
About this item
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline
Boiling Pt:245 °C (1013 hPa)
Melting Pt:220…222 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 245 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 220…222 °C
:Flash Pt:
:Flash Pt:
:M.W. 115.13 g/mol
Merck Index:12,07963
:L(-)-Proline
:245 °C (1013 hPa)
Formula:C₅H₉NO₂
MW:115.13 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00064318
CAS Number:147-85-3
Customers Who Bought This Also Bought