Acetonitrile ≥99.9% for LC-MS, Pierce™
Catalog #: CAPI85188
Supplier: Thermo Scientific
CAS Number: 75-05-8
Specifications
- Size:1 L
- Cat. no.:CAPI85188
- Supplier No.:85188
Specifications
Specification Test Results
- Acetonitrile ≥99.9% for LC-MS, Pierce™:
Specification Test Results
About this item
Synonyms: Methyl cyanide,Ethyl nitrile,Cyanomethane,Ethanenitrile
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:2 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 81.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –45.7 °C
:Flash Pt:
:Flash Pt: 2 °C (closed cup)
:M.W. 41.05 g/mol
:Acetonitrile
MW:41.05 g/mol
MDL Number:MFCD00001878
UN:1648
:3
:II
:2 °C
Formula:H₃CCN
CAS Number:75-05-8
Customers Who Bought This Also Bought