Trifluoroacetic acid ≥99.5% for LC-MS, Pierce™
Catalog #: CAPI85183
Supplier: Thermo Scientific
CAS Number: 76-05-1
Specifications
- Size:50 ml
- Cat. no.:CAPI85183
- Supplier No.:85183
Specifications
Specification Test Results
- Trifluoroacetic acid ≥99.5% for LC-MS, Pierce™:
Specification Test Results
About this item
Synonyms: TFA
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
ADR:8,I
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt: 71.8 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –15 °C
:Flash Pt:
:Flash Pt:
:M.W. 114.02 g/mol
Formula:C₂HF₃O₂
MW:114.02 g/mol
UN:2699
:8
:I
:12,09812
MDL Number:MFCD00004169
CAS Number:76-05-1
Customers Who Bought This Also Bought