Trifluoroacetic acid 0.1% (v/v) in acetonitrile for LC-MS, Pierce™
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: TFA
Formula:C₂HF₃O₂
MW:114.02 g/mol
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.5351 g/cm³ (20 °C)
MDL Number:MFCD00004169
CAS Number:76-05-1
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 71.8 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –15 °C
:Flash Pt:
:Flash Pt:
:M.W. 114.02 g/mol
UN:1648
:3
:II
:12,09812
Specification Test Results
- in acetonitrile:
Specification Test Results
Related Information
Customers Who Bought This Also Bought