Pierce™ UltraLink™ Iodoacetyl Resin, Affinity Purification Resins, Thermo Scientific
Catalog #: CAPI53155
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:UltraLink™ Iodoacetyl Resin
- Size:10 mL
- Storage conditions:Storage: Upon receipt store product at 4˚C, protected from light.
- Cat. no.:CAPI53155
- Supplier No.:53155
Specifications
About this item
UltraLink™ Iodoacetyl Resin is a durable, porous resin that is activated to enable efficient and direct covalent immoblization of peptides and other ligands through their sulfhydryl groups (-SH) for use in affinity purification procedures.
- High coupling efficiency and capacity
- Specific and leak-proof coupling chemistry
- Durable, high-performance resin
- Prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification
The bead structure and efficiently coupling chemistry of the Iodoacetyl-activated UltraLink™ support results in high protein binding capacity, high linear flow rates, low non specific binding and overall superior performance in affinity chromatography. UltraLink™ resins are ideal for medium pressure applications, such as FPLC.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought