To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pierce™ Clear Milk blocking buffer (10X)
- Size:100 mL
- Cat. no.:CAPI37587
- Supplier No.:37587
Specifications
About this item
Thermo Scientific Pierce Clear Milk Blocking Buffer (10X) is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
- Kit stable for 1 year stored at 4 °C
- Convenient and saves storage space
- Easy to use with ready-to-dilute solution
Clear Milk Blocking Buffer (10X) is a proprietary pre-formulated milk solution for blocking excess nonspecific binding sites, reducing background in Western blotting applications and diluting antibodies when used with nitrocellulose and PVDF membranes. Pierce Clear Milk Blocking Buffer (10X) provides lower background, enhanced sensitivity, extended shelf life and reproducible results compared to homemade buffers based on dissolved non-fat dry milk.