Order Entry
ContactUsLinkComponent
Rapid ELISA Mouse mAb Isotyping Kit, Thermo Scientific
Rapid ELISA Mouse mAb Isotyping Kit, Thermo Scientific
Catalog #: CAPI37503
Supplier:  Thermo Scientific
CAS Number:  
Rapid ELISA Mouse mAb Isotyping Kit, Thermo Scientific
Catalog #: CAPI37503
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Reactivity:
    Mouse
  • Size:
    60 test kit
  • Cat. no.:
    CAPI37503
  • Supplier No.:
    37503

Specifications

About this item

The Thermo Scientific Pierce Rapid ELISA Mouse mAb Isotyping Kit provides antigen-independent determination of mouse monoclonal antibody isotype using a microplate method that enables multiple samples to be tested in about one hour.

Features of the Rapid ELISA Mouse mAb Isotyping Kit:

Fast—determine antibody isotype in approx. 1 hour; considerably faster than traditional ELISA kits
Convenient—eight-well strip format allows partial use of the plate; use one strip (column) for each sample (12 samples per plate)
Specific—characterize antibodies for six different subclasses and two different light-chain types
Sensitive—accurate characterization with samples containing at least 3 ng/mL of test antibody
Flexible—kit is compatible with hybridoma cell culture supernatant, ascites fluid or purified antibodies
Cost effective—cost per assay is less than with single-use assay strips or cassettes
No special equipment needed—assess results visually or measure quantitatively using an ordinary ELISA plate reader (450nm)
Complete and easy to use—includes precoated plates, detecting antibody, buffers, and TMB substrate and stop solutions

This fast assay uses ELISA strip-plates that are pre-coated in different wells with anti-mouse heavy-chain capture antibodies (anti-IgG1, IgG2a, IgG2b, IgG3, IgA and IgM) or anti-mouse light-chain capture antibody (kappa or lambda). This approach eliminates the need to purify and immobilize an antigen to determine immunoglobulin isotype (antibody subclass and light chain identity). A mouse monoclonal antibody sample applied to the wells can be isotyped within one hour. Results are evaluated qualitatively by visual inspection or quantitatively by measuring the absorbance at 450nm.

Ordering information: For Research Use Only. Not for use in diagnostic procedures. Sufficient For: 60 isotype determinations (8 wells per test sample)
Pre-coated 96-well Isotyping Plates, 5 plates
Goat Anti-Mouse IgG+IgA+IgM-HRP Conjugate, 30 mL
TMB Substrate, 55 mL
Stop Solution, 55 mL
30X Wash Buffer, 200 mL
TBS Buffer Packs (each makes 500 mL), 2 packs
Store at 4˚C.
Customers Who Bought This Also Bought